missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-17/IL-17A Recombinant Protein, (CHO-expressed)

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16171
Molecular Weight 15 kDa (monomer)
Name IL-17/IL-17A
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7956ML025 R&D Systems
RDS7956ML025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $298.51
7956ML025CF R&D Systems
RDS7956ML025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $298.51
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ala26-Ala158
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16171
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16171
IL-17/IL-17A
25μg
Mouse