Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-17/IL-17A Recombinant Protein

$292.21 - $292.21

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16171
Molecular Weight 15.5 kDa (monomer)
Name IL-17/IL-17A
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

421ML025

 
r&d systems
RDS421ML025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $292.21

421ML025CF

 
r&d systems
RDS421ML025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $292.21
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16171
Molecular Weight 15.5 kDa (monomer)
Name IL-17/IL-17A
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Thr22-Ala158
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 16171
Regulatory Status: RUO