missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-12/IL-23 p40 Homodimer Recombinant Protein

$454.80 - $454.80

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16160
Molecular Weight 35.8 kDa (monomer)
Name IL-12/IL-23 p40 Homodimer
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
499ML025CF R&D Systems
RDS499ML025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.80
499ML025 R&D Systems
RDS499ML025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.80
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Met23-Ser335
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 35.8 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16160
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
35.8 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16160
IL-12/IL-23 p40 Homodimer
25μg
Mouse