missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-11 Recombinant Protein

$363.69 - $1,166.69

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16156
Molecular Weight 19 kDa
Name IL-11
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
418ML005CF R&D Systems
RDS418ML005CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. 5μg Each for $363.69
418ML025CF R&D Systems
RDS418ML025CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. 25μg Each for $1,166.69
418ML025 R&D Systems
RDS418ML025
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein. 25μg Each for $1,166.69
418ML005 R&D Systems
RDS418ML005
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein. 5μg Each for $363.69
Specifications

Description

Source: E. coli-derived, Gly23-Leu199, with an N-terminal Met and Pro25-Leu199
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Entrez Gene ID: 16156
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
16156
IL-11
Yes