missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-11 R alpha Fc Chimera Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16157
Molecular Weight 65 kDa (monomer)
Name IL-11 R alpha Fc Chimera
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
490IR200CF R&D Systems
RDS490IR200CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser22-Gln367
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 65 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16157
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
65 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16157
IL-11 R alpha Fc Chimera
200μg
Mouse