missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-10 Recombinant Protein

$324.58 - $1,062.40

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16153
Molecular Weight 18 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
417ML005CF R&D Systems
RDS417ML005CF
Supplied as a 0.2μm filtered solution in Tris and NaCl. 5μg Each for $324.58
417ML005 R&D Systems
RDS417ML005
Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein. 5μg Each for $324.58
417ML025 R&D Systems
RDS417ML025
Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein. 25μg Each for $1,062.40
417ML025CF R&D Systems
RDS417ML025CF
Supplied as a 0.2μm filtered solution in Tris and NaCl. 25μg Each for $1,062.40
Specifications

Description

Source: E. coli-derived, Ser19-Ser178
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Supplied as a 0.2μm filtered solution in Tris and NaCl.
Entrez Gene ID: 16153
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
16153
IL-10
Yes