missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-10 R alpha Recombinant Protein, Thr24-Ile241

$518.14 - $518.14

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16154
Molecular Weight 25.5 kDa
Name IL-10 R alpha
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7704MR025CF R&D Systems
RDS7704MR025CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $518.14
7704MR025 R&D Systems
RDS7704MR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $518.14
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (stably transfected)-derived, Thr24-Ile241
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Entrez Gene ID: 16154
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
25.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16154
IL-10 R alpha
25μg
Mouse