missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-10 R alpha Recombinant Protein

$533.15 - $533.15

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16154
Molecular Weight 24 kDa
Name IL-10 R alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
474MR025CF R&D Systems
RDS474MR025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $533.15
474MR025 R&D Systems
RDS474MR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $533.15
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Thr24-Asn238 (Lys188Arg and Glu222Gly)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16154
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
24 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16154
IL-10 R alpha
25μg
Mouse