missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-10 (Cys167Tyr) Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16153
Molecular Weight 18.8 kDa (monomer)
Name IL-10 (Cys167Tyr)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1023ML010 R&D Systems
RDS1023ML010
Lyophilized from a 0.2μm filtered solution in Sodium Phosphate with BSA as a carrier protein. Each for $376.73
1023ML010CF R&D Systems
RDS1023ML010CF
Lyophilized from a 0.2μm filtered solution in Sodium Phosphate. Each for $376.73
Specifications

Description

Source: E. coli-derived, Ser19-Ser178 (Cys167Tyr), with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.8 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Phosphate.
Entrez Gene ID: 16153
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.8 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16153
IL-10 (Cys167Tyr)
10μg
Mouse