missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-1 Rrp2/IL-1 R6 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 107527
Molecular Weight 36.8 kDa
Name IL-1 Rrp2/IL-1 R6
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2354RP100 R&D Systems
RDS2354RP100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
2354RP100CF R&D Systems
RDS2354RP100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Asp22-Arg338 (Ala158Val), with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 107527
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
36.8 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
107527
IL-1 Rrp2/IL-1 R6
100μg
Mouse