missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-1 alpha/IL-1F1 Recombinant Protein

$337.62 - $1,062.40

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16175
Molecular Weight 18 kDa
Name IL-1 alpha/IL-1F1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
400ML005 R&D Systems
RDS400ML005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $337.62
400ML025 R&D Systems
RDS400ML025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $1,062.40
400ML005CF R&D Systems
RDS400ML005CF
Supplied as a 0.2 mm filtered solution in PBS. 5μg Each for $337.62
400ML025CF R&D Systems
RDS400ML025CF
Supplied as a 0.2 mm filtered solution in PBS. 25μg Each for $1,062.40
Specifications

Description

Source: E. coli-derived, Ser6-Ser161
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Supplied as a 0.2mm filtered solution in PBS.
Entrez Gene ID: 16175
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
16175
IL-1 alpha/IL-1F1
Yes