missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IGF-I R Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 104.5 kDa (single chain), 80.7 kDa ( alpha subunit) & 23.2 kDa (beta subunit)
Name IGF-I R
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6630GR025 R&D Systems
RDS6630GR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
6630GR025CF R&D Systems
RDS6630GR025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-His936, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 104.5 kDa (single chain), 80.7 kDa ( alpha subunit) & 23.2 kDa (beta subunit)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IGF-I R
25μg
Mouse
104.5 kDa (single chain), 80.7 kDa ( alpha subunit) & 23.2 kDa (beta subunit)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes