Please login to your online account to display your discounted pricing

R&D Systems Mouse IFN-gamma Recombinant Protein

$529.55 - $529.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 15978
Molecular Weight 15.6 kDa
Name IFN-gamma
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

485MI100CF

 
r&d systems
RDS485MI100CF
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol, TCEP and Tween™ 80 with Trehalose. Each for $529.55

485MI100

 
r&d systems
RDS485MI100
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol, TCEP and Tween™ 80 with BSA as a carrier protein. Each for $529.55
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 15978
Molecular Weight 15.6 kDa
Name IFN-gamma
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 100μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, His23-Cys155, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol, TCEP and Tween™ 80 with Trehalose.
Entrez Gene ID: 15978
Regulatory Status: RUO