Please login to your online account to display your discounted pricing

R&D Systems Mouse Hip Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 15245
Name Hip
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity    

1568HP050

 
r&d systems
RDS1568HP050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 76 kDa & 73 kDa Each for $419.81

1568HP050CF

 
r&d systems
RDS1568HP050CF
Lyophilized from a 0.2μm filtered solution in PBS. 76 kDa and 73 kDa Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 15245
Name Hip
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Lys24-Arg678 & Asp52-Arg678, both with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 76 kDa and 73 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 15245
Regulatory Status: RUO