Please login to your online account to display your discounted pricing

R&D Systems Mouse Gremlin Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 23892
Molecular Weight 19.6 kDa
Name Gremlin
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

956GR050

 
r&d systems
RDS956GR050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $441.90

956GR050CF

 
r&d systems
RDS956GR050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $441.90
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 23892
Molecular Weight 19.6 kDa
Name Gremlin
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Lys25-Asp184, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 23892
Regulatory Status: RUO