Please login to your online account to display your discounted pricing

R&D Systems Mouse Glypican 5 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 103978
Molecular Weight 59.1 kDa
Name Glypican 5
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2689G5050

 
r&d systems
RDS2689G5050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

2689G5050CF

 
r&d systems
RDS2689G5050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 103978
Molecular Weight 59.1 kDa
Name Glypican 5
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Gly26-Gly549, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 59.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 103978
Regulatory Status: RUO