missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse GITR Ligand/TNFSF18 aa 47-173 Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 240873
Molecular Weight 16 kDa
Name GITR Ligand/TNFSF18 aa 47-173
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2177GL025 R&D Systems
RDS2177GL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $441.90
2177GL025CF R&D Systems
RDS2177GL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $441.90
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Thr47-Ser173, with an N-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 240873
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
16 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
240873
GITR Ligand/TNFSF18 aa 47-173
25μg
Mouse