missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Gas6 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14456
Molecular Weight 62 kDa
Name Gas6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
986GS025CF R&D Systems
RDS986GS025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
986GS025 R&D Systems
RDS986GS025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Asp115-Pro673, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 62 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 14456
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
62 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
14456
Gas6
25μg
Mouse