R&D Systems Mouse Galectin-7 Recombinant Protein
Manufacturer:  R&D Systems RDS1304GA050
 View more versions of this product
Catalog No. 1304GA050
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein.
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
16858
Galectin-7
50μg
Unlabeled