missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Galectin-7 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16858
Molecular Weight 15 kDa
Name Galectin-7
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1304GA050CF R&D Systems
RDS1304GA050CF
Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol. Each for $428.87
1304GA050 R&D Systems
RDS1304GA050
Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: E. coli-derived, Ser2-Phe136
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
Entrez Gene ID: 16858
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16858
Galectin-7
50μg
Mouse