Please login to your online account to display your discounted pricing

R&D Systems Mouse Galectin-7 Recombinant Protein


Manufacturer: r&d systems  RDS1304GA050CF

 View more versions of this product

Catalog No. 1304GA050CF

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 15 kDa
Name Galectin-7
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Source Mouse
Formulation Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
Gene ID (Entrez) 16858
Recombinant Yes
Conjugate Unlabeled