Please login to your online account to display your discounted pricing

R&D Systems Mouse Galectin-3 Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16854
Molecular Weight 27.3 kDa
Name Galectin-3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1197GA050

 
r&d systems
RDS1197GA050
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16854
Molecular Weight 27.3 kDa
Name Galectin-3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Ala2-Ile264
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 27.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
Entrez Gene ID: 16854
Regulatory Status: RUO