missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse G-CSF Recombinant Protein

$272.44 - $846.01

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12985
Molecular Weight 19 kDa
Name G-CSF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
414CS005 R&D Systems
RDS414CS005
Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20 with BSA as a carrier protein. 5μg Each for $272.44
414CS025 R&D Systems
RDS414CS025
Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20 with BSA as a carrier protein. 25μg Each for $846.01
414CS005CF R&D Systems
RDS414CS005CF
Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20. 5μg Each for $272.44
414CS025CF R&D Systems
RDS414CS025CF
Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20. 25μg Each for $846.01
Specifications

Description

Source: E. coli-derived, Val31-Ala208, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20.
Entrez Gene ID: 12985
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
12985
G-CSF
Yes