missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Follistatin 288 (FS-288) Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14313
Molecular Weight 31 kDa
Name Follistatin 288 (FS-288)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
769FS025 R&D Systems
RDS769FS025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87
769FS025CF R&D Systems
RDS769FS025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gly30-Asn317, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 31 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 14313
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
31 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
14313
Follistatin 288 (FS-288)
25μg
Mouse