missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Flt-3 Ligand Recombinant Protein

$194.23 - $624.40

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14256
Molecular Weight 18 kDa
Name Flt-3 Ligand
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
427FL005 R&D Systems
RDS427FL005
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 5μg Each for $194.23
427FL005CF R&D Systems
RDS427FL005CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 5μg Each for $194.23
427FL025 R&D Systems
RDS427FL025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 25μg Each for $624.40
427FL025CF R&D Systems
RDS427FL025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 25μg Each for $624.40
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gly27-Arg188
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 14256
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
14256
Flt-3 Ligand
Yes