missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse FGF basic Recombinant Protein

$337.62 - $337.62

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14173
Molecular Weight 16.2 kDa
Name FGF basic
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3139FB025 R&D Systems
RDS3139FB025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein. Each for $337.62
3139FB025CF R&D Systems
RDS3139FB025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. Each for $337.62
Specifications

Description

Source: E. coli-derived, Ala11-Ser154
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Entrez Gene ID: 14173
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
16.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
14173
FGF basic
25μg
Mouse