missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse FGF acidic Recombinant Protein

$233.33 - $233.33

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14164
Molecular Weight 16 kDa
Name FGF acidic
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4686FA025CF R&D Systems
RDS4686FA025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. Each for $233.33
4686FA025 R&D Systems
RDS4686FA025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein. Each for $233.33
Specifications

Description

Source: E. coli-derived, Phe16-Asp155, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Entrez Gene ID: 14164
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
16 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
14164
FGF acidic
25μg
Mouse