missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse FGF-8 c Isoform Recombinant Protein

$328.11 - $328.11

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14179
Molecular Weight 28 kDa
Name FGF-8 c Isoform
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
424FC025 R&D Systems
RDS424FC025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $328.11
424FC025CF R&D Systems
RDS424FC025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $328.11
Specifications

Description

Source: E. coli-derived, Gln23-Arg268, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 28 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 14179
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
28 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
14179
FGF-8 c Isoform
25μg
Mouse