missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse FGF-6 Recombinant Protein

$328.11 - $328.11

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14177
Molecular Weight 18.8 kDa
Name FGF-6
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5750F6025 R&D Systems
RDS5750F6025
Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTT with BSA as a carrier protein. Each for $328.11
5750F6025CF R&D Systems
RDS5750F6025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTT. Each for $328.11
Specifications

Description

Source: E. coli-derived, Pro39-Ile208
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTT.
Entrez Gene ID: 14177
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.8 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
14177
FGF-6
25μg
Mouse