Please login to your online account to display your discounted pricing

R&D Systems Mouse FGF-4 Recombinant Protein

$330.49 - $330.49

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14175
Molecular Weight 19.0 kDa
Name FGF-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

5846F4025

 
r&d systems
RDS5846F4025
Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTT with BSA as a carrier protein. Each for $330.49

5846F4025CF

 
r&d systems
RDS5846F4025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTT and Trehalose. Each for $330.49
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14175
Molecular Weight 19.0 kDa
Name FGF-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Ala30-Leu202
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.0 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Ammonium Sulfate, EDTA, DTTand Trehalose.
Entrez Gene ID: 14175
Regulatory Status: RUO