missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse FGF-10 Recombinant Protein

$337.62 - $337.62

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14165
Molecular Weight 17.1 kDa
Name FGF-10
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6224FG025 R&D Systems
RDS6224FG025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein. Each for $337.62
6224FG025CF R&D Systems
RDS6224FG025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. Each for $337.62
Specifications

Description

Source: E. coli-derived, Ser62-Thr209
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Entrez Gene ID: 14165
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
14165
FGF-10
25μg
Mouse