missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Fas Ligand/TNFSF6 Recombinant Protein

$467.46 - $467.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14103
Name Fas Ligand/TNFSF6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
526SA050 R&D Systems
RDS526SA050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 18 kDa (monomer) Each for $467.46
526SA050CF R&D Systems
RDS526SA050CF
Lyophilized from a 0.2μm filtered solution in PBS. 18 kDa Each for $467.46
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Pro132-Leu279, with an N-terminal Met and 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 14103
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
Fas Ligand/TNFSF6
50μg
Mouse
14103
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes