missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Erythropoietin Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 13856
Molecular Weight 18.6 kDa
Name Erythropoietin
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
959ME010 R&D Systems
RDS959ME010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
959ME010CF R&D Systems
RDS959ME010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala27-Arg192
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 13856
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.6 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
13856
Erythropoietin
10μg
Mouse