missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse DLL4 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 54485
Molecular Weight 55.6 kDa
Name DLL4
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1389D4050 R&D Systems
RDS1389D4050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
1389D4050CF R&D Systems
RDS1389D4050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser28-Pro525, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 55.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 54485
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
55.6 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
54485
DLL4
50μg
Mouse