missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Dkk-4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 234130
Molecular Weight 23 kDa
Name Dkk-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3105DK010 R&D Systems
RDS3105DK010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
3105DK010CF R&D Systems
RDS3105DK010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu19-Ile221 (Arg67Ser and Arg70Ser), with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 23 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 234130
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
23 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
234130
Dkk-4
10μg
Mouse