missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Dkk-2 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 56811
Molecular Weight 27.1 kDa
Name Dkk-2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2435DK010CF R&D Systems
RDS2435DK010CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $416.79
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ser26-Ile259, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 27.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Entrez Gene ID: 56811
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
27.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
56811
Dkk-2
10μg
Mouse