missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Dkk-1 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 13380
Molecular Weight 26.1 kDa
Name Dkk-1
Purity 95%, by SDS-PAGE with silver staining.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5897DK010 R&D Systems
RDS5897DK010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
5897DK010CF R&D Systems
RDS5897DK010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser30-His272
Purity: 95%, by SDS-PAGE with silver staining.
Molecular Weight: 26.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 13380
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
26.1 kDa
95%, by SDS-PAGE with silver staining.
Yes
13380
Dkk-1
10μg
Mouse