missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Dkk-1 (Histidine-tagged) Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 13380
Molecular Weight 27.6 kDa
Name Dkk-1 (Histidine-tagged)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1765DK010CF R&D Systems
RDS1765DK010CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $416.79
1765DK010 R&D Systems
RDS1765DK010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser30-His272, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 27.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Entrez Gene ID: 13380
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
27.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
13380
Dkk-1 (Histidine-tagged)
10μg
Mouse