Please login to your online account to display your discounted pricing

R&D Systems Mouse DcTRAIL R2/TNFRSF22 Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 79202
Molecular Weight 16.5 kDa
Name DcTRAIL R2/TNFRSF22
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1816DT050

 
r&d systems
RDS1816DT050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94

1816DT050CF

 
r&d systems
RDS1816DT050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 79202
Molecular Weight 16.5 kDa
Name DcTRAIL R2/TNFRSF22
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Ala39-Arg175 & Met40-Arg175, both with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 79202
Regulatory Status: RUO