missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL7/Thymus Chemokine-1 aa 48-109 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 57349
Molecular Weight 6.8 kDa
Name CXCL7/Thymus Chemokine-1 aa 48-109
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1091CK025CF R&D Systems
RDS1091CK025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
1091CK025 R&D Systems
RDS1091CK025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: E. coli-derived, Ile48-Ile109
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 6.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 57349
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
6.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
57349
CXCL7/Thymus Chemokine-1 aa 48-109
25μg
Mouse