Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL7/Thymus Chemokine-1 aa 40-113 Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 57349
Molecular Weight 8.2 kDa
Name CXCL7/Thymus Chemokine-1 aa 40-113
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

793CK025

 
r&d systems
RDS793CK025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

793CK025CF

 
r&d systems
RDS793CK025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 57349
Molecular Weight 8.2 kDa
Name CXCL7/Thymus Chemokine-1 aa 40-113
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Lys40-Tyr113
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 57349
Regulatory Status: RUO