missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL2/GRO beta/MIP-2/CINC-3 Recombinant Protein

$311.55 - $559.23

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20310
Molecular Weight 8 kDa
Name CXCL2/GRO beta/MIP-2/CINC-3
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
452M2010 R&D Systems
RDS452M2010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $311.55
452M2050 R&D Systems
RDS452M2050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $559.23
452M2050CF R&D Systems
RDS452M2050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $559.23
452M2010CF R&D Systems
RDS452M2010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $311.55
Specifications

Description

Source: E. coli-derived, Ala28-Asn100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20310
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
20310
CXCL2/GRO beta/MIP-2/CINC-3
Yes