missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL17/VCC-1 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 232983
Molecular Weight 10.3 kDa
Name CXCL17/VCC-1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4270DM025CF R&D Systems
RDS4270DM025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $298.51
4270DM025 R&D Systems
RDS4270DM025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $298.51
Specifications

Description

Source: E. coli-derived, Ser23-Leu119
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 232983
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
10.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
232983
CXCL17/VCC-1
25μg
Mouse