Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL16 Extracellular Domain Recombinant Protein, Asn27-Trp201


Manufacturer: r&d systems  RDS1165CX025

 View more versions of this product

Catalog No. 1165CX025

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 19.9 kDa
Name CXCL16 Extracellular Domain
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Mouse
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 66102
Quantity 25μg