Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL16 Extracellular Domain Recombinant Protein, Asn27-Trp201

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 66102
Molecular Weight 19.9 kDa
Name CXCL16 Extracellular Domain
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1165CX025CF

 
r&d systems
RDS1165CX025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $298.51

1165CX025

 
r&d systems
RDS1165CX025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $298.51
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 66102
Molecular Weight 19.9 kDa
Name CXCL16 Extracellular Domain
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Asn27-Trp201, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 66102
Regulatory Status: RUO