missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL16 Chemokine Domain Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 66102
Molecular Weight 9.9 kDa
Name CXCL16 Chemokine Domain
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
503CX025 R&D Systems
RDS503CX025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $298.51
503CX025CF R&D Systems
RDS503CX025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $298.51
Specifications

Description

Source: E. coli-derived, Asn27-Pro114
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 66102
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
9.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
66102
CXCL16 Chemokine Domain
25μg
Mouse