missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL12/SDF-1 alpha Recombinant Protein

$298.51 - $559.23

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20315
Molecular Weight 8 kDa
Name CXCL12/SDF-1 alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
460SD010CF R&D Systems
RDS460SD010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $298.51
460SD010 R&D Systems
RDS460SD010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $298.51
460SD050 R&D Systems
RDS460SD050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $559.23
460SD050CF R&D Systems
RDS460SD050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $559.23
Specifications

Description

Source: E. coli-derived, Lys22-Lys89
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20315
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
20315
CXCL12/SDF-1 alpha
Yes