missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL11/I-TAC Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 56066
Molecular Weight 9.1 kDa
Name CXCL11/I-TAC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
572MC025 R&D Systems
RDS572MC025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51
572MC025CF R&D Systems
RDS572MC025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51
Specifications

Description

Source: E. coli-derived, Phe22-Met100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 56066
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
9.1 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
56066
CXCL11/I-TAC
25μg
Mouse