missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL1/GRO alpha/KC/CINC-1 aa 29-96 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14825
Molecular Weight 7.5 kDa
Name CXCL1/GRO alpha/KC/CINC-1 aa 29-96
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1395KC025CF R&D Systems
RDS1395KC025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $389.76
1395KC025 R&D Systems
RDS1395KC025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $389.76
Specifications

Description

Source: E. coli-derived, Asn29-Lys96
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 14825
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
14825
CXCL1/GRO alpha/KC/CINC-1 aa 29-96
25μg
Mouse