Please login to your online account to display your discounted pricing

R&D Systems Mouse CXCL1/GRO alpha/KC/CINC-1 aa 20-96 Recombinant Protein

$298.51 - $559.23

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14825
Molecular Weight 8 kDa
Name CXCL1/GRO alpha/KC/CINC-1 aa 20-96
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

453KC010CF

 
r&d systems
RDS453KC010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $298.51

453KC010

 
r&d systems
RDS453KC010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $298.51

453KC050

 
r&d systems
RDS453KC050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $559.23

453KC050CF

 
r&d systems
RDS453KC050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $559.23
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 14825
Molecular Weight 8 kDa
Name CXCL1/GRO alpha/KC/CINC-1 aa 20-96
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Mouse

Source: E. coli-derived, Arg20-Lys96
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 14825
Regulatory Status: RUO